Learn More
Waters Corp Waters Corporation Target Plate, 384-Well, 16×24 Grid, ANSI & SBS Standard
Catalog No. 50793247 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 384-well target plate is a critical laboratory tool for scientific and biomedical applications, featuring 384 smaller wells in a 16×24 grid for ultra-high-throughput experiments. It is specifically designed for use with the MALDI SYNAPT XS system.
  • Features 384 smaller wells in a 16×24 grid.
  • Adheres to ANSI and SBS standards.
  • Constructed from polypropylene (PP) for chemical resistance.
  • Suitable for LC autosamplers and liquid handling robots.
  • Compatible with various sealing options, including cap mats, heat seals, and adhesive seals.
  • Ideal for ultra-high-throughput applications, assays, and sample storage.
  • Facilitates automated liquid handling and workflow integration.
  • Recommended for high-throughput LC and LC–MS work.
  • Utilized in research, diagnostics, and drug discovery.
Catalog No. 50-793-247 Supplier Waters Corp Supplier No. M880356BD1S
This 384-well target plate is a critical laboratory tool for scientific and biomedical applications, featuring 384 smaller wells in a 16×24 grid for ultra-high-throughput experiments. It is specifically designed for use with the MALDI SYNAPT XS system.
  • Features 384 smaller wells in a 16×24 grid.
  • Adheres to ANSI and SBS standards.
  • Constructed from polypropylene (PP) for chemical resistance.
  • Suitable for LC autosamplers and liquid handling robots.
  • Compatible with various sealing options, including cap mats, heat seals, and adhesive seals.
  • Ideal for ultra-high-throughput applications, assays, and sample storage.
  • Facilitates automated liquid handling and workflow integration.
  • Recommended for high-throughput LC and LC–MS work.
  • Utilized in research, diagnostics, and drug discovery.