Learn More
Medchemexpress LLC Targocil | 1200443-21-5 | 99.3% | 475.95 | 200 MG
SDP
Catalog No. 5000388153
Encompass_Preferred
Targocil is a bacteriostatic inhibitor of wall teichoic acid (WTA) biosynthesis. It is effective against both methicillin-susceptible S. aureus (MSSA) and methicillin-resistant S. aureus (MRSA) strains, with MIC90s of 2 μg/mL for both. It shows excellent activity against S. aureus isolates from bacterial keratitis.
  • Bacteriostatic inhibitor of wall teichoic acid (WTA) biosynthesis
  • Inhibits growth of methicillin-susceptible S. aureus (MSSA)
  • Inhibits growth of methicillin-resistant S. aureus (MRSA)
  • Effective against S. aureus strains Newman, MW2, MG2375, and MG2389
  • Low toxicity for HCECs at 5 μg/mL after 24 h exposure
Catalog No. 50-003-88153 Supplier Medchemexpress LLC Supplier No. HY18702200MG
Targocil is a bacteriostatic inhibitor of wall teichoic acid (WTA) biosynthesis. It is effective against both methicillin-susceptible S. aureus (MSSA) and methicillin-resistant S. aureus (MRSA) strains, with MIC90s of 2 μg/mL for both. It shows excellent activity against S. aureus isolates from bacterial keratitis.
  • Bacteriostatic inhibitor of wall teichoic acid (WTA) biosynthesis
  • Inhibits growth of methicillin-susceptible S. aureus (MSSA)
  • Inhibits growth of methicillin-resistant S. aureus (MRSA)
  • Effective against S. aureus strains Newman, MW2, MG2375, and MG2389
  • Low toxicity for HCECs at 5 μg/mL after 24 h exposure