Learn More
Medchemexpress LLC Taribavirin hydrochloride | 40372-00-7 | MFCD00876899 | 99.9% | 279.68 g/mol | C8H14ClN5O4 | 100 MG
SDP
Catalog No. 5000240231
Encompass_Preferred
Taribavirin hydrochloride is the hydrochloride salt of taribavirin, an orally available prodrug of ribavirin supplied for research use. Provided as a solid with high purity for in vitro and preclinical studies, it is soluble in DMSO and requires sealed, low-temperature storage for long-term stability.
  • High purity suitable for research applications.
  • Solid form for accurate weighing and formulation.
  • Soluble in DMSO at approximately 100 mg/mL with ultrasonic aid.
  • Stable when stored sealed at 4°C; in solution store at -80°C for long-term.
  • Designed to concentrate in the liver for antiviral research applications.
Catalog No. 50-002-40231 Supplier Medchemexpress LLC Supplier No. HY10545A100MG
Taribavirin hydrochloride is the hydrochloride salt of taribavirin, an orally available prodrug of ribavirin supplied for research use. Provided as a solid with high purity for in vitro and preclinical studies, it is soluble in DMSO and requires sealed, low-temperature storage for long-term stability.
  • High purity suitable for research applications.
  • Solid form for accurate weighing and formulation.
  • Soluble in DMSO at approximately 100 mg/mL with ultrasonic aid.
  • Stable when stored sealed at 4°C; in solution store at -80°C for long-term.
  • Designed to concentrate in the liver for antiviral research applications.