Learn More
Medchemexpress LLC Taribavirin hydrochloride | 40372-00-7 | MFCD00876899 | 99.9% | 279.68 g/mol | C8H14ClN5O4 | 25 MG
SDP
Catalog No. 5000240521
Encompass_Preferred
Taribavirin hydrochloride is an orally active inosine monophosphate dehydrogenase (IMPDH) inhibitor and ribavirin prodrug used in antiviral research. It shows activity against hepatitis C virus and influenza in preclinical studies and is supplied in high-purity solid and solution forms for laboratory use.
  • Orally active IMPDH inhibitor
  • Demonstrated antiviral activity vs HCV and influenza in preclinical studies
  • Designed to concentrate in the liver to target infected hepatocytes
  • Available as solid vials and as a DMSO solution for assays
  • High reported purity suitable for research applications
  • Supplied in small quantities convenient for laboratory experiments
Catalog No. 50-002-40521 Supplier Medchemexpress LLC Supplier No. HY10545A25MG
Taribavirin hydrochloride is an orally active inosine monophosphate dehydrogenase (IMPDH) inhibitor and ribavirin prodrug used in antiviral research. It shows activity against hepatitis C virus and influenza in preclinical studies and is supplied in high-purity solid and solution forms for laboratory use.
  • Orally active IMPDH inhibitor
  • Demonstrated antiviral activity vs HCV and influenza in preclinical studies
  • Designed to concentrate in the liver to target infected hepatocytes
  • Available as solid vials and as a DMSO solution for assays
  • High reported purity suitable for research applications
  • Supplied in small quantities convenient for laboratory experiments