Learn More
Selleck Chemical LLC Tariquidar 10mg 206873-63-4 XR9576
SDP
Supplier:  Selleck Chemical LLC S802810MG
Encompass_Preferred
Tariquidar (XR9576) is a potent and selective noncompetitive inhibitor of P-glycoprotein with Kd of 5.1 nM in CHrB30 cell line, reverses drug resistance in MDR cell Lines. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0783015