Learn More
Selleck Chemical LLC Tariquidar
SDP
Supplier:  Selleck Chemical LLC S802810MM/1ML
Encompass_Preferred
Tariquidar is a potent and selective noncompetitive inhibitor of P-glycoprotein with Kd of 5 1 nM in CHrB30 cell line reverses drug resistance in MDR cell Lines Phase 3
Catalog No. 50-136-5577