Learn More
American Radiolabeled Chemicals Inc TARIQUIDAR 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 175650UC
Encompass_Preferred
Tariquidar-methyl-3H 50uCi in ethanol, 1 mCi/ml, S.A. 70-87 Ci/mmol, M.W. 646.73, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-189