Learn More
Medchemexpress LLC HY-10550 5mg , Tariquidar CAS:206873-63-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105505MG
Encompass_Preferred
HY-10550 5mg Tariquidar CAS: 206873-63-4 Tariquidar (XR9576) is a potent and specific inhibitor of P-glycoprotein (P-gp) with the high affinity (Kd=5.1 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9620