Learn More
Medchemexpress LLC HY-10550 100mg , Tariquidar XR9576 CAS:206873-63-4 Purity:98%
SDP
Catalog No. 501493403
Encompass_Preferred
Medchemexpress, HY-10550 100mg Tariquidar XR9576 CAS:206873-63-4 Formula:C38H38N4O6 Kd: 5.1 nM (P-gp) Purity:98% Tariquidar is a potent and specific inhibitor of P-glycoprotein ( P-gp ) with the high affinity ( K d =5.1±0.9 nM). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3403 Supplier Medchemexpress LLC Supplier No. HY10550100MG
Medchemexpress, HY-10550 100mg Tariquidar XR9576 CAS:206873-63-4 Formula:C38H38N4O6 Kd: 5.1 nM (P-gp) Purity:98% Tariquidar is a potent and specific inhibitor of P-glycoprotein ( P-gp ) with the high affinity ( K d =5.1±0.9 nM). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.