Learn More
Medchemexpress LLC HY-10550A 10mg Medchemexpress, Tariquidar (methanesulfonate, hydrate) CAS:625375-83-9 Purity:>98%
SDP
Catalog No. 501968273
Encompass_Preferred
Medchemexpress, HY-10550A 10mg Tariquidar (methanesulfonate, hydrate) CAS:625375-83-9 Tariquidar methanesulfonate, hydrate (XR9576 methanesulfonate, hydrate) is a potent and specific inhibitor of P-glycoprotein (P-gp) with a Kd of 5.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8273 Supplier Medchemexpress LLC Supplier No. HY10550A10MG
Medchemexpress, HY-10550A 10mg Tariquidar (methanesulfonate, hydrate) CAS:625375-83-9 Tariquidar methanesulfonate, hydrate (XR9576 methanesulfonate, hydrate) is a potent and specific inhibitor of P-glycoprotein (P-gp) with a Kd of 5.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.