Learn More
Medchemexpress LLC Tarloxotinib bromide | 1636180-98-7 | 98.75% | 681.77 | 1 ML
SDP
Catalog No. 5000386975
Encompass_Preferred
Tarloxotinib bromide is an irreversible EGFR/HER2 inhibitor that is efficiently metabolized under hypoxia in human NSCLC cell lines. Cellular anti-proliferative and receptor phosphorylation assays show a 14-80 fold reduction of activity relative to TKI, demonstrating greater activity compared with erlotinib. In vivo studies show that treatment with clinically relevant doses leads to tumor regression and durable inhibition of WT EGFR tumor phosphorylation, and can regress WT EGFR NSCLC tumor models H125 and H1648.
  • Irreversible EGFR/HER2 inhibitor
  • Efficiently metabolized under hypoxia
  • Greater activity compared with erlotinib
  • Associated with tumor regression and durable inhibition of WT EGFR tumor phosphorylation
  • Can regress WT EGFR NSCLC tumor models H125 and H1648
Catalog No. 50-003-86975 Supplier Medchemexpress LLC Supplier No. HY1763210MM/1ML
Tarloxotinib bromide is an irreversible EGFR/HER2 inhibitor that is efficiently metabolized under hypoxia in human NSCLC cell lines. Cellular anti-proliferative and receptor phosphorylation assays show a 14-80 fold reduction of activity relative to TKI, demonstrating greater activity compared with erlotinib. In vivo studies show that treatment with clinically relevant doses leads to tumor regression and durable inhibition of WT EGFR tumor phosphorylation, and can regress WT EGFR NSCLC tumor models H125 and H1648.
  • Irreversible EGFR/HER2 inhibitor
  • Efficiently metabolized under hypoxia
  • Greater activity compared with erlotinib
  • Associated with tumor regression and durable inhibition of WT EGFR tumor phosphorylation
  • Can regress WT EGFR NSCLC tumor models H125 and H1648