Learn More
Medchemexpress LLC Tarloxotinib bromide | 1636180-98-7 | 98.8% | 681.77 | 1 MG
SDP
Catalog No. 5000386977
Encompass_Preferred
Tarloxotinib bromide (TH-4000) is an irreversible EGFR/HER2 inhibitor and a hypoxia-activated irreversible EGFR-TKI. This product appears as a light yellow to yellow solid with a high purity of 98.75%.
  • Irreversible EGFR/HER2 inhibitor
  • Hypoxia-activated irreversible EGFR-TKI
  • High purity
  • Appears as a light yellow to yellow solid
  • Soluble in DMSO
  • Suitable for in vitro and in vivo studies
Catalog No. 50-003-86977 Supplier Medchemexpress LLC Supplier No. HY176321MG
Tarloxotinib bromide (TH-4000) is an irreversible EGFR/HER2 inhibitor and a hypoxia-activated irreversible EGFR-TKI. This product appears as a light yellow to yellow solid with a high purity of 98.75%.
  • Irreversible EGFR/HER2 inhibitor
  • Hypoxia-activated irreversible EGFR-TKI
  • High purity
  • Appears as a light yellow to yellow solid
  • Soluble in DMSO
  • Suitable for in vitro and in vivo studies