Learn More
TARGETMOL CHEMICALS INC Tarloxotinib bromide 1MG
SDP
Catalog No. 502876970
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tarloxotinib bromide (TH-4000) is an irreversible inhibitor of EGFR/HER2. Purity 97.15%
Catalog No. 50-287-6970 Supplier TARGETMOL CHEMICALS INC Supplier No. T130881MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tarloxotinib bromide (TH-4000) is an irreversible inhibitor of EGFR/HER2. Purity 97.15%