Learn More
Grainger Buyers Products Electric Tarp Gear Motor, 12V, 600W, 90:1, 1 ea.
Catalog No. 502786564
Encompass_Preferred
This 600 W electric tarp gear motor is designed for reliable performance in demanding dump applications and electric tarp systems. Featuring a durable, weather-resistant enclosure, the unit utilizes a 90:1 gear ratio to provide robust torque for smooth operation. This bi-directional, 12 V DC motor is engineered for long-term durability and includes a chrome-plated cover and essential mounting hardware.
  • 600 W power output for efficient tarp operation
  • Bi-directional rotation
  • Durable bronze alloy gear construction
  • Weatherproof enclosure
  • 90:1 gear ratio
  • Fits universal 3 and 4 bolt mounting patterns
  • 3-year warranty
Catalog No. 50-278-6564 Supplier Grainger Supplier No. 19A794
This 600 W electric tarp gear motor is designed for reliable performance in demanding dump applications and electric tarp systems. Featuring a durable, weather-resistant enclosure, the unit utilizes a 90:1 gear ratio to provide robust torque for smooth operation. This bi-directional, 12 V DC motor is engineered for long-term durability and includes a chrome-plated cover and essential mounting hardware.
  • 600 W power output for efficient tarp operation
  • Bi-directional rotation
  • Durable bronze alloy gear construction
  • Weatherproof enclosure
  • 90:1 gear ratio
  • Fits universal 3 and 4 bolt mounting patterns
  • 3-year warranty