Learn More
Grainger Buyers Products Electric Tarp Gear Motor, 12V, 900W, 1 ea
Catalog No. 502785789
Encompass_Preferred
This electric tarp gear motor is designed to automate the process of covering dump truck beds, utilizing a high-speed 900 W, 12V motor to efficiently deploy and retract tarps. The system features a 90:1 gear ratio, delivering 40 RPM for reliable operation and consistent performance during daily fleet use.
  • Features a durable bronze alloy gear for extended service life
  • Includes a protective, weatherproof enclosure with a chrome-plated cover
  • Designed with 5 mounting holes to accommodate both 3-bolt and 4-bolt installation patterns
  • Provides bi-directional rotation for versatile tarp deployment
  • Water-resistant design suited for exterior vehicle environments
Catalog No. 50-278-5789 Supplier Grainger Supplier No. 19A795
This electric tarp gear motor is designed to automate the process of covering dump truck beds, utilizing a high-speed 900 W, 12V motor to efficiently deploy and retract tarps. The system features a 90:1 gear ratio, delivering 40 RPM for reliable operation and consistent performance during daily fleet use.
  • Features a durable bronze alloy gear for extended service life
  • Includes a protective, weatherproof enclosure with a chrome-plated cover
  • Designed with 5 mounting holes to accommodate both 3-bolt and 4-bolt installation patterns
  • Provides bi-directional rotation for versatile tarp deployment
  • Water-resistant design suited for exterior vehicle environments