Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Tartan™ Filament Tape

Catalog No. 19072102 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
55 m x 48 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-102 55 m x 48 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-102 Supplier 3M™ Supplier No. 7000028888
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tartan™ Filament Tape is a general purpose clear fiberglass-reinforced tape with a synthetic rubber resin adhesive, ideal for light duty strapping, bundling and reinforcing.

  • Light-duty high strength tape for utility bundling, reinforcing and pallet unitizing applications
  • Synthetic rubber adhesive bonds easily to most fiberboard, plastics and metal surfaces
  • Transparency provides clear visibility of box graphics, printing and illustrations
  • Tape resists nicks, abrasions, moisture and scuffing
  • Glass yarn filaments add strength for box closing, bundling and more
TRUSTED_SUSTAINABILITY

Specifications

Product Type Filament Tape
For Use With (Application) Box Closing, Bundling, Palletizing, Reinforcement, Strapping, Utility
Adhesive Type Synthetic Rubber
Color Clear
Dimensions (L x W) 55 m x 48 mm
Material Synthetic Rubber
Model 8934
Quantity 24 Rolls/Cs.
Thickness (English) 4 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.