Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Tartan™ Pistol Grip Box Sealing Tape Dispenser

Catalog No. 19072040 Shop All 3M Products
Change view
Click to view available options
Quantity:
24/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-040 24/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-040 Supplier 3M™ Supplier No. 7000131573
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tartan™ Pistol Grip Box Sealing Tape Dispenser is an affordable, portable, hand-held tape dispenser designed with a pistol-grip design for easy application.

  • One-hand, pistol-grip design allows for easy application
  • Tack, roll, buff and tear in one fluid motion
  • Plastic and metal construction creates a rugged and durable tool
  • Lightweight, rugged cobstruction creates a durable, yet portable tool for sealing, combining, reinforcing and bundling
  • Adjustable, hub-mounted tape brake offers control of tension and delivery
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sealing Tape Dispenser
For Use With (Application) Automated Carton Sealing, Box Closure, General and Primary Packaging
Color Black
Model HB903
Quantity 24/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.