Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tartaric Acid, 2% (w/v), Ricca Chemical
SDP

Catalog No. 837032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8370-32 1 L Natural Poly Bottle
8370-16 500 mL Natural Poly Bottle
R83700004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 8370-32 Supplier Ricca Chemical Company Supplier No. 837032
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) Mixture
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

CAS Min % 96.0, 1.92
Color Colorless to Yellow
CAS Max % 100.0, 2.0
Concentration 2% (w/v)
Percent Purity 2% w/v
Grade Laboratory
Quantity 1 L
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Tartaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.