Learn More
Selleck Chemical LLC Tartaric acid S2997-100mg
SDP
Catalog No. 503109603
Encompass_Preferred
Tartaric acid (2 3-Dihydroxysuccinic acid) is a white crystalline organic acid isolated from many plants particularly tamarinds and grapes and is used as an antioxidant and an additive agent to give a sour taste
Catalog No. 50-310-9603 Supplier Selleck Chemical LLC Supplier No. S2997100MG
Tartaric acid (2 3-Dihydroxysuccinic acid) is a white crystalline organic acid isolated from many plants particularly tamarinds and grapes and is used as an antioxidant and an additive agent to give a sour taste