Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tartaric Acid AR Granular, Macron Fine Chemicals™

Catalog No. MK2312500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-2312-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-2312-500 Supplier Avantor Supplier No. 231204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 87-69-4
Quantity 500 g
Packaging POLYSTORMOR
Physical Form Granular
Molecular Formula C4H6O6
MDL Number MFCD00064207
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES OC(C(O)C(O)=O)C(O)=O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.09
PubChem CID 444305
ChEBI CHEBI:15671
Formula Weight 150.09
Chemical Name or Material Tartaric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.