Learn More
Medchemexpress LLC Tartaric acid disodium dihydrate | 6106-24-7 | 98.0% | 1 ML
SDP
Catalog No. 5000410439
Encompass_Preferred
Tartaric acid disodium dihydrate is an orally active weak organic acid that can be isolated from grapes. It has vasodilatory and antihypertensive effects. It can be used as flavorings and antioxidants in a range of foods and beverages, and also in laser frequency doubling and optical limiting applications.
  • Orally active weak organic acid
  • Can be isolated from grapes
  • Has vasodilatory and antihypertensive effects
  • Can be used as flavorings and antioxidants in food and beverages
  • Can be used in laser frequency doubling and optical limiting applications
Catalog No. 50-004-10439 Supplier Medchemexpress LLC Supplier No. HYD085010MM/1ML
Tartaric acid disodium dihydrate is an orally active weak organic acid that can be isolated from grapes. It has vasodilatory and antihypertensive effects. It can be used as flavorings and antioxidants in a range of foods and beverages, and also in laser frequency doubling and optical limiting applications.
  • Orally active weak organic acid
  • Can be isolated from grapes
  • Has vasodilatory and antihypertensive effects
  • Can be used as flavorings and antioxidants in food and beverages
  • Can be used in laser frequency doubling and optical limiting applications