Learn More
Hach Company Tartrate Buffer Solution, 1 L (Lachat reagent for TKN FIA)
Catalog No. NC2112245 Shop All Hach Company Products
Encompass
Tartrate buffer solution formulated for total Kjeldahl nitrogen (TKN) analysis by flow-injection analysis (FIA). Supplied as a 1 L prepared reagent. Hazardous shipping may apply; a Safety Data Sheet (SDS) is available.
  • 1 L volume.
  • Prepared reagent for TKN analysis using flow-injection analysis (FIA).
  • May be classified as hazardous for shipping; safety data sheet available.
  • Suitable for use with automated flow-injection analysis systems.
Catalog No. NC2112245 Supplier Hach Company Supplier No. 52009
Tartrate buffer solution formulated for total Kjeldahl nitrogen (TKN) analysis by flow-injection analysis (FIA). Supplied as a 1 L prepared reagent. Hazardous shipping may apply; a Safety Data Sheet (SDS) is available.
  • 1 L volume.
  • Prepared reagent for TKN analysis using flow-injection analysis (FIA).
  • May be classified as hazardous for shipping; safety data sheet available.
  • Suitable for use with automated flow-injection analysis systems.