Learn More
Electron Microscopy Sciences Tartrazine, 1.5% Aqueous Sol 500 ML
SDP
Catalog No. 5031266
Encompass_Preferred
For bacterial stain. See AFIP Manual of Histologic Staining Methods; ed. G. Luna, McGraw-Hill Publications, 3rd. ed.; New York, c. 1968, p.224 or Brown, R.C. and Hopps, H.C., Geographic Pathology Division Armed Forces Institute of Pathology, Washington, D.C. 20305
Catalog No. 50-312-66 Supplier Electron Microscopy Sciences Supplier No. 2610606
For bacterial stain. See AFIP Manual of Histologic Staining Methods; ed. G. Luna, McGraw-Hill Publications, 3rd. ed.; New York, c. 1968, p.224 or Brown, R.C. and Hopps, H.C., Geographic Pathology Division Armed Forces Institute of Pathology, Washington, D.C. 20305