Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tartrazine, 89%, pure

Catalog No. AC191895000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191895000 500 g Glass bottle
AC191891000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191895000 Supplier Thermo Scientific Chemicals Supplier No. 191895000
  • $215.50
    $158.65 / Each of 1

    Save $56.85

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1934-21-0 | C16H9N4Na3O9S2 | 534.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1934-21-0
Molecular Formula C16H9N4Na3O9S2
Molecular Weight (g/mol) 534.35
MDL Number MFCD00148908
InChI Key UJMBCXLDXJUMFB-UHFFFAOYSA-K
Synonym tartrazine, yellow 5, aizen tartrazine, trisodium salt, food yellow 4, tartrazine FD&C yellow #5, c.i. acid yellow 23, atul tartrazine, erio tartrazine, kako tartrazine
PubChem CID 164825
IUPAC Name trisodium;5-oxo-1-(4-sulfonatophenyl)-4-[(4-sulfonatophenyl)diazenyl]-4H-pyrazole-3-carboxylate
SMILES C1=CC(=CC=C1N=NC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)[O-])C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Decomposition Information >300°C
Melting Point >300.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 89%
Packaging Glass bottle
Quantity 500 g
Beilstein 25, IV, 1757
Merck Index 15, 9206
Solubility Information Solubility in water: 260g/L water (30°C)
Formula Weight 534.35
Percent Purity 89%
Grade Pure
Insoluble Matter 0.2% max. (in water)
Physical Form Powder
Chemical Name or Material Tartrazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 217-699-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.