Learn More
Medchemexpress LLC Tartrazine (acid yellow 23; FD&C yellow no. 5) | 1934-21-0 | 99.8% | 500 MG
SDP
Catalog No. 5000410488
Encompass_Preferred
Tartrazine, also known as Acid Yellow 23 or FD&C Yellow No. 5, is an orally active azo acid dye presented as an orange-yellow powder that is soluble in water, turning yellow upon dissolution. This dye is predominantly utilized as a synthetic lemon yellow food coloring agent, recognized for its stability. Research indicates that Tartrazine can induce mitochondria-mediated apoptosis and is associated with neurodevelopmental toxicity, cytotoxicity, and genotoxicity. This product is strictly for research purposes and is not intended for patient use.
  • Orally active azo acid dye
  • Orange-yellow powder form
  • Water-soluble, producing a yellow solution
  • Used as a synthetic lemon yellow food coloring
  • Considered a stable colorant
  • Induces mitochondria-mediated apoptosis
  • Associated with neurodevelopmental toxicity
  • Exhibits cytotoxicity and genotoxicity
  • For research use only
Catalog No. 50-004-10488 Supplier Medchemexpress LLC Supplier No. HYD0257500MG
Tartrazine, also known as Acid Yellow 23 or FD&C Yellow No. 5, is an orally active azo acid dye presented as an orange-yellow powder that is soluble in water, turning yellow upon dissolution. This dye is predominantly utilized as a synthetic lemon yellow food coloring agent, recognized for its stability. Research indicates that Tartrazine can induce mitochondria-mediated apoptosis and is associated with neurodevelopmental toxicity, cytotoxicity, and genotoxicity. This product is strictly for research purposes and is not intended for patient use.
  • Orally active azo acid dye
  • Orange-yellow powder form
  • Water-soluble, producing a yellow solution
  • Used as a synthetic lemon yellow food coloring
  • Considered a stable colorant
  • Induces mitochondria-mediated apoptosis
  • Associated with neurodevelopmental toxicity
  • Exhibits cytotoxicity and genotoxicity
  • For research use only