Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tartrazine, Sigma-Aldrich

Catalog No. 01258982 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-982 100 g
01-258-876 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-982 Supplier MilliporeSigma Supelco Supplier No. T0388100G
Add to Cart
Add to Cart

Tartrazine may be used as a reference standard in the determination of tartrazine in food samples using ion-pair liquid chromatography coupled with photodiode-array and electrospray mass spectrometry detector.

The application of Tartrazine is seen in the phloxine-tartrazine procedure for cellular inclusions such as viral aggregates and eosinophil granules. It may be used as a substitute for orange G in a variant of Mallory's trichrome, and in a methyl violet-tartrazine mixture to stain cryptosporidia in faecal smears and mucosal scrapings.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥85%
Quantity 100 g
Product Type Tartrazine Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.