Learn More
Tartrazine Solution, 250 mL, G-Biosciences
SCI_ED
Catalog No. S001136
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
S001136 250 mL
1 options
Catalog No. S001136 Supplier G-Biosciences Supplier No. 7862242
Add to Cart
Add to Cart
Tartrazine Solution is used as a counterstain for a wide range of histological procedures.
  • It is used to make living tissues transparent in mice to observe internal organs and blood vessels.
Specifications
Quantity 250 mL
Product Type Tartrazine Solution