Learn More
Biomatik Corporation Tartrazine (TZ) ELISA Kit
Supplier:  Biomatik Corporation EKU11457
Encompass_Preferred
Tartrazine (TZ) ELISA Kit for detecting TZ in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 617.3-50,000pg/ml. Sensitivity: 284.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Trisodium 1-(4-sulfonatophenyl)-4-(4-sulfonatophenylazo)-5-pyrazolone-3-carboxylate). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8140