Learn More
TARGETMOL CHEMICALS INC TAS-103 dihydrochloride 10MG
SDP
Catalog No. 502877237
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAS-103 dihydrochloride (BMS-247615 dihydrochloride) is a novel anticancer agent targeting both topoisomerase (Topo) I and Topo II. Purity 97.78%
Catalog No. 50-287-7237 Supplier TARGETMOL CHEMICALS INC Supplier No. T730910MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAS-103 dihydrochloride (BMS-247615 dihydrochloride) is a novel anticancer agent targeting both topoisomerase (Topo) I and Topo II. Purity 97.78%