Learn More
Medchemexpress LLC TAS-103 dihydrochloride | 174634-09-4 | 98.5% | 406.31 | 50 MG
SDP
Catalog No. 5000351813
Encompass_Preferred
TAS-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II, used for cancer research.
  • Dual inhibitor of DNA topoisomerase I/II.
  • Active on CCRF-CEM cells with an IC50 value of 5 nM.
  • Significantly increases levels of topo IIα FITC immunofluorescence in individual CCRF-CEM cells.
  • Highly cytotoxic to Lewis lung carcinoma (LLC) cells.
  • Inhibits the viability of HeLa cells with an IC50 of 40 nM.
  • Disrupts signal recognition particle (SRP) complex formation and induces destabilization of SRP14 and SRP19.
  • Causes significant tumor growth suppression in mice bearing Lewis lung carcinoma (LLC) cells without obvious body weight loss.
  • Liposomal TAS-103 is more active than free TAS-103 in vivo.
Catalog No. 50-003-51813 Supplier Medchemexpress LLC Supplier No. HY13758A50MG
TAS-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II, used for cancer research.
  • Dual inhibitor of DNA topoisomerase I/II.
  • Active on CCRF-CEM cells with an IC50 value of 5 nM.
  • Significantly increases levels of topo IIα FITC immunofluorescence in individual CCRF-CEM cells.
  • Highly cytotoxic to Lewis lung carcinoma (LLC) cells.
  • Inhibits the viability of HeLa cells with an IC50 of 40 nM.
  • Disrupts signal recognition particle (SRP) complex formation and induces destabilization of SRP14 and SRP19.
  • Causes significant tumor growth suppression in mice bearing Lewis lung carcinoma (LLC) cells without obvious body weight loss.
  • Liposomal TAS-103 is more active than free TAS-103 in vivo.