Learn More
Medchemexpress LLC Tas-103 dihydrochloride (BMS-247615 dihydrochloride) | 174634-09-4 | 98.5% | 406.31 | 1 ML
SDP
Catalog No. 5000348503
Encompass_Preferred
Tas-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II, utilized for cancer research. It demonstrates activity on CCRF-CEM cells with an IC50 value of 5 nM and is highly cytotoxic to Lewis lung carcinoma (LLC) cells. It also inhibits the viability of HeLa cells.
  • Dual inhibitor of DNA topoisomerase I/II.
  • Demonstrates activity on CCRF-CEM cells with an IC50 value of 5 nM.
  • Elevates levels of topo IIα FITC immunofluorescence in individual CCRF-CEM cells.
  • Highly cytotoxic to Lewis lung carcinoma (LLC) cells.
  • Inhibits viability of HeLa cells, exhibiting an IC50 of 40 nM.
  • Disrupts signal recognition particle (SRP) complex formation.
  • Suppresses tumor growth in mice bearing Lewis lung carcinoma (LLC) cells in vivo.
Catalog No. 50-003-48503 Supplier Medchemexpress LLC Supplier No. HY13758A10MM/1ML
Tas-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II, utilized for cancer research. It demonstrates activity on CCRF-CEM cells with an IC50 value of 5 nM and is highly cytotoxic to Lewis lung carcinoma (LLC) cells. It also inhibits the viability of HeLa cells.
  • Dual inhibitor of DNA topoisomerase I/II.
  • Demonstrates activity on CCRF-CEM cells with an IC50 value of 5 nM.
  • Elevates levels of topo IIα FITC immunofluorescence in individual CCRF-CEM cells.
  • Highly cytotoxic to Lewis lung carcinoma (LLC) cells.
  • Inhibits viability of HeLa cells, exhibiting an IC50 of 40 nM.
  • Disrupts signal recognition particle (SRP) complex formation.
  • Suppresses tumor growth in mice bearing Lewis lung carcinoma (LLC) cells in vivo.