Learn More
Medchemexpress LLC HY-124062 5mg , TAS-114 CAS:1198221-21-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1240625MG
Encompass_Preferred
HY-124062 5mg TAS-114 CAS: 1198221-21-4 TAS-114 is a dual dUTPase/dihydropyrimidine dehydrogenase (DPD) inhibitor, can improving the therapeutic efficacy of fluoropyrimidine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0625