Learn More
Medchemexpress LLC TAS-119 | 1453099-83-6 | 98.0% | 506.36 | 1 ML
SDP
Catalog No. 5000350399
Encompass_Preferred
TAS-119 is a potent, selective, and orally active Aurora A inhibitor. It demonstrates high selectivity for Aurora A over other protein kinases and exhibits potent antitumor activities.
  • Potent and selective Aurora A inhibitor with an IC50 of 1.0 nM.
  • Shows high selectivity for Aurora A over Aurora B (IC50 of 95 nM).
  • Enhances the antiproliferative effect of paclitaxel in various human cancer cell lines, including paclitaxel-resistant cells.
  • Dose-dependently enhances cell growth inhibition by taxanes.
  • Induces mitotic accumulation predominantly in tumor cells.
Catalog No. 50-003-50399 Supplier Medchemexpress LLC Supplier No. HY13737710MM/1ML
TAS-119 is a potent, selective, and orally active Aurora A inhibitor. It demonstrates high selectivity for Aurora A over other protein kinases and exhibits potent antitumor activities.
  • Potent and selective Aurora A inhibitor with an IC50 of 1.0 nM.
  • Shows high selectivity for Aurora A over Aurora B (IC50 of 95 nM).
  • Enhances the antiproliferative effect of paclitaxel in various human cancer cell lines, including paclitaxel-resistant cells.
  • Dose-dependently enhances cell growth inhibition by taxanes.
  • Induces mitotic accumulation predominantly in tumor cells.