Learn More
Medchemexpress LLC Tas-119 | 1453099-83-6 | 98.0% | 506.36 | 100 MG
SDP
Catalog No. 5000351357
Encompass_Preferred
TAS-119 is a potent, selective, and orally active Aurora A inhibitor with an IC50 of 1.0 nM. It demonstrates high selectivity over Aurora B and exhibits potent antitumor activities. It enhances the antiproliferative effect of Paclitaxel in various human cancer cell lines.
  • Potent and selective Aurora A inhibitor
  • High selectivity over Aurora B
  • Exhibits potent antitumor activities
  • Enhances antiproliferative effect of Paclitaxel
  • Induces mitotic accumulation in tumor cells
  • Orally active
Catalog No. 50-003-51357 Supplier Medchemexpress LLC Supplier No. HY137377100MG
TAS-119 is a potent, selective, and orally active Aurora A inhibitor with an IC50 of 1.0 nM. It demonstrates high selectivity over Aurora B and exhibits potent antitumor activities. It enhances the antiproliferative effect of Paclitaxel in various human cancer cell lines.
  • Potent and selective Aurora A inhibitor
  • High selectivity over Aurora B
  • Exhibits potent antitumor activities
  • Enhances antiproliferative effect of Paclitaxel
  • Induces mitotic accumulation in tumor cells
  • Orally active