Learn More
Medchemexpress LLC TAS-119 | 1453099-83-6 | 98.0% | 506.36 | 25 MG
SDP
Catalog No. 5000352127
Encompass_Preferred
TAS-119 is a potent, selective, and orally active Aurora A inhibitor. It exhibits an IC50 of 1.0 nM for Aurora A and 95 nM for Aurora B. This compound has demonstrated potent antitumor activities, enhancing the antiproliferative effect of Paclitaxel in various human cancer cell lines, including drug-resistant ones.
  • Potent, selective, and orally active Aurora A inhibitor
  • IC50 of 1.0 nM for Aurora A
  • High selectivity over Aurora B (IC50 of 95 nM)
  • Demonstrates potent antitumor activities
  • Enhances antiproliferative effect of Paclitaxel
  • Induces mitotic accumulation in tumor cells
  • For research use only
Catalog No. 50-003-52127 Supplier Medchemexpress LLC Supplier No. 25MGHY137377
Add to Cart
Add to Cart
TAS-119 is a potent, selective, and orally active Aurora A inhibitor. It exhibits an IC50 of 1.0 nM for Aurora A and 95 nM for Aurora B. This compound has demonstrated potent antitumor activities, enhancing the antiproliferative effect of Paclitaxel in various human cancer cell lines, including drug-resistant ones.
  • Potent, selective, and orally active Aurora A inhibitor
  • IC50 of 1.0 nM for Aurora A
  • High selectivity over Aurora B (IC50 of 95 nM)
  • Demonstrates potent antitumor activities
  • Enhances antiproliferative effect of Paclitaxel
  • Induces mitotic accumulation in tumor cells
  • For research use only