Learn More
Medchemexpress LLC HY-137377 5mg Medchemexpress, TAS-119 CAS:1453099-83-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1373775MG
Encompass_Preferred
Medchemexpress, HY-137377 5mg TAS-119 CAS:1453099-83-6 TAS-119 is a potent, selective and orally active Aurora A inhibitor with an IC50 of 1.0 nM. TAS-119 shows high selectivity for Aurora A over other protein kinases, including Aurora B (IC50 of 95 nM). TAS-119 has potent antitumor activites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1487