Learn More
Medchemexpress LLC HY-18965 5mg Medchemexpress, TAS-301 CAS:193620-69-8 Purity:>98%
SDP
Catalog No. 502082102
Encompass_Preferred
Medchemexpress, HY-18965 5mg TAS-301 CAS:193620-69-8 TAS-301 is an inhibitor of smooth muscle cell migration and proliferation, and inhibits PKC activation induced by PDGF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2102 Supplier Medchemexpress LLC Supplier No. HY189655MG
Medchemexpress, HY-18965 5mg TAS-301 CAS:193620-69-8 TAS-301 is an inhibitor of smooth muscle cell migration and proliferation, and inhibits PKC activation induced by PDGF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.