Learn More
Medchemexpress LLC HY-111553 10mg Medchemexpress, TAS0728 CAS:2088323-16-2 Purity:>98%
SDP
Catalog No. 501968918
Encompass_Preferred
Medchemexpress, HY-111553 10mg TAS0728 CAS:2088323-16-2 TAS0728 is a potent, selective, oral active, irreversible and covalent-binding HER2 inhibitor, binds to HER2 at C805, inhibits its kinase activity, with an IC50 of 13 nM. TAS0728 shows IC50s of 4.9, 8.5, 31, 65, 33, 25, 86 and 36 nM for BMX, HER4, BLK, EGFR, JAK3, SLK, LOK and human HER2, respectively. TAS0728 also inhibits the phosphorylation of HER2, HER3, and downstream effectors, shows no obvious effect on EGFR. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8918 Supplier Medchemexpress LLC Supplier No. HY11155310MG
Medchemexpress, HY-111553 10mg TAS0728 CAS:2088323-16-2 TAS0728 is a potent, selective, oral active, irreversible and covalent-binding HER2 inhibitor, binds to HER2 at C805, inhibits its kinase activity, with an IC50 of 13 nM. TAS0728 shows IC50s of 4.9, 8.5, 31, 65, 33, 25, 86 and 36 nM for BMX, HER4, BLK, EGFR, JAK3, SLK, LOK and human HER2, respectively. TAS0728 also inhibits the phosphorylation of HER2, HER3, and downstream effectors, shows no obvious effect on EGFR. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.