Learn More
ABclonal Technology TAS2R10 Rabbit pAb
SDP
Supplier:  ABclonal Technology A151561000UL
Encompass_Preferred
This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13.
Catalog No. 50-001-08175