Learn More
Medchemexpress LLC HY-128586 5mg , TAS4464 CAS:1848959-10-3 Purity:>98%
SDP
Catalog No. 502260662
Encompass_Preferred
HY-128586 5mg TAS4464 CAS: 1848959-10-3 TAS4464 is a highly potent and selective inhibitor of NEDD8 activating enzyme (NAE), with an IC50 of 0.955 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0662 Supplier Medchemexpress LLC Supplier No. HY1285865MG
HY-128586 5mg TAS4464 CAS: 1848959-10-3 TAS4464 is a highly potent and selective inhibitor of NEDD8 activating enzyme (NAE), with an IC50 of 0.955 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.