Learn More
Selleck Chemical LLC TAS4464 S8849-5MG
SDP
Catalog No. 5000025343
Encompass_Preferred
TAS4464 is a potent and highly selective NEDD8 activating enzyme (NAE) inhibitor with IC50 of 0 955 nM TAS4464 exhibits superior antitumor activity with prolonged target inhibition
Catalog No. 50-000-25343 Supplier Selleck Chemical LLC Supplier No. S88495MG
TAS4464 is a potent and highly selective NEDD8 activating enzyme (NAE) inhibitor with IC50 of 0 955 nM TAS4464 exhibits superior antitumor activity with prolonged target inhibition