Learn More
Medchemexpress LLC Tas4464 hydrochloride | 1848959-11-4 | 98.9% | 1 ML
SDP
Catalog No. 5000339165
Encompass_Preferred
TAS4464 hydrochloride is a long-acting, highly selective covalent inhibitor that targets the NEDD8-activating enzyme (NAE) with an IC50 of 0.955 nM. It blocks the neddylation pathway, leading to the accumulation of CRL ubiquitin ligase substrates and subsequent induction of tumor cell apoptosis. This compound exhibits broad-spectrum antiproliferative and cytotoxic antitumor activity against various hematologic and solid tumor cell lines, as well as patient-derived tumor cells, while possessing a wide therapeutic window without obvious toxicity.
  • Highly selective covalent inhibitor of NEDD8-activating enzyme (NAE)
  • Blocks the neddylation pathway
  • Induces tumor cell apoptosis
  • Exhibits broad-spectrum antitumor activity
  • Possesses a wide therapeutic window
  • Suitable for research in hematologic malignancies and solid tumors
Catalog No. 50-003-39165 Supplier Medchemexpress LLC Supplier No. HY128586A10MM/1ML
TAS4464 hydrochloride is a long-acting, highly selective covalent inhibitor that targets the NEDD8-activating enzyme (NAE) with an IC50 of 0.955 nM. It blocks the neddylation pathway, leading to the accumulation of CRL ubiquitin ligase substrates and subsequent induction of tumor cell apoptosis. This compound exhibits broad-spectrum antiproliferative and cytotoxic antitumor activity against various hematologic and solid tumor cell lines, as well as patient-derived tumor cells, while possessing a wide therapeutic window without obvious toxicity.
  • Highly selective covalent inhibitor of NEDD8-activating enzyme (NAE)
  • Blocks the neddylation pathway
  • Induces tumor cell apoptosis
  • Exhibits broad-spectrum antitumor activity
  • Possesses a wide therapeutic window
  • Suitable for research in hematologic malignancies and solid tumors