Learn More
Medchemexpress LLC Tas4464 (hydrochloride) | 1848959-11-4 | 98.9% | 25 MG
SDP
Catalog No. 5000341631
Encompass_Preferred
TAS4464 hydrochloride is a long-acting, highly selective covalent inhibitor targeting NEDD8-activating enzyme (NAE) with an IC50 of 0.955 nM. It exhibits antiproliferative and cytotoxic effects with broad-spectrum antitumor activity against various hematologic and solid tumor cell lines and patient-derived tumor cells. This compound has a wide therapeutic window without obvious toxicity and is suitable for research in hematologic malignancies and solid tumors.
  • Highly potent and selective NAE inhibitor
  • Demonstrates broad-spectrum antitumor activity against various cancer models
  • Induces tumor cell apoptosis
  • Increases protein expression levels of CRL ubiquitin ligase substrates
  • Inhibits migratory capacity of solid tumor cell lines
  • Achieves complete tumor regression in various animal models
  • Prolongs survival and reduces tumor burden in lymphoma models
  • Possesses a wide therapeutic window without obvious toxicity
Catalog No. 50-003-41631 Supplier Medchemexpress LLC Supplier No. 25MGHY128586A
TAS4464 hydrochloride is a long-acting, highly selective covalent inhibitor targeting NEDD8-activating enzyme (NAE) with an IC50 of 0.955 nM. It exhibits antiproliferative and cytotoxic effects with broad-spectrum antitumor activity against various hematologic and solid tumor cell lines and patient-derived tumor cells. This compound has a wide therapeutic window without obvious toxicity and is suitable for research in hematologic malignancies and solid tumors.
  • Highly potent and selective NAE inhibitor
  • Demonstrates broad-spectrum antitumor activity against various cancer models
  • Induces tumor cell apoptosis
  • Increases protein expression levels of CRL ubiquitin ligase substrates
  • Inhibits migratory capacity of solid tumor cell lines
  • Achieves complete tumor regression in various animal models
  • Prolongs survival and reduces tumor burden in lymphoma models
  • Possesses a wide therapeutic window without obvious toxicity