Learn More
Medchemexpress LLC Zipalertinib | 1661854-97-2 | 99.6% | 396.44 | 10 MG
SDP
Catalog No. 501969075
Encompass_Preferred
Zipalertinib (TAS6417; CLN-081) is a highly effective, orally active, and pan-mutation-selective EGFR tyrosine kinase inhibitor. It features a unique scaffold that fits into the ATP-binding site of the EGFR hinge region, with IC50 values ranging from 1.1-8.0 nM. It inhibits EGFR phosphorylation and downstream molecules in NSCLC cell lines and causes persistent tumor regression in EGFR exon 20 insertion-driven tumor models.
  • Highly effective, orally active, and pan-mutation-selective EGFR tyrosine kinase inhibitor
  • Inhibits EGFR phosphorylation and downstream molecules
  • Causes caspase activation
  • Robust against common EGFR mutations and potent against cells with EGFR-T790M
  • Inhibits mutant EGFR in tumors but not wild-type EGFR in skin
  • Promotes cell growth inhibition and apoptosis
  • Prolongs survival in animal models of lung cancer
Catalog No. 50-196-9075 Supplier Medchemexpress LLC Supplier No. HY11229910MG
Zipalertinib (TAS6417; CLN-081) is a highly effective, orally active, and pan-mutation-selective EGFR tyrosine kinase inhibitor. It features a unique scaffold that fits into the ATP-binding site of the EGFR hinge region, with IC50 values ranging from 1.1-8.0 nM. It inhibits EGFR phosphorylation and downstream molecules in NSCLC cell lines and causes persistent tumor regression in EGFR exon 20 insertion-driven tumor models.
  • Highly effective, orally active, and pan-mutation-selective EGFR tyrosine kinase inhibitor
  • Inhibits EGFR phosphorylation and downstream molecules
  • Causes caspase activation
  • Robust against common EGFR mutations and potent against cells with EGFR-T790M
  • Inhibits mutant EGFR in tumors but not wild-type EGFR in skin
  • Promotes cell growth inhibition and apoptosis
  • Prolongs survival in animal models of lung cancer