Learn More
MSC Tasco Earmuffs, 28 dB NRR, Three Position Band, 1 ea
Catalog No. 5000782107
Encompass
These earmuffs offer effective hearing protection for various applications. They feature a 28 dB Noise Reduction Rating and a three-position band for comfortable and versatile wear.
  • Earmuff design for noise reduction
  • Padded headband for enhanced comfort
  • Gray cup color
  • High visibility for safety
  • Meets S3.19-1974 standards
Catalog No. 50-007-82107 Supplier MSC Supplier No. 02947521
These earmuffs offer effective hearing protection for various applications. They feature a 28 dB Noise Reduction Rating and a three-position band for comfortable and versatile wear.
  • Earmuff design for noise reduction
  • Padded headband for enhanced comfort
  • Gray cup color
  • High visibility for safety
  • Meets S3.19-1974 standards