Learn More
MSC Tasco Mesh Screen, Black, 6 in. High, 1 ea
Catalog No. 5000677623
Encompass
This mesh screen face shield provides protection for various applications. It is designed for compatibility with specific headgear systems.
  • Mesh screen
  • Black color
  • Measures 6 in. × 15-1/2 in.
  • Epoxy coating
  • No lens shade
  • Compatible with T-Lock Ratchet Style Headgear and Woodsman System
Catalog No. 50-006-77623 Supplier MSC Supplier No. 02015238
This mesh screen face shield provides protection for various applications. It is designed for compatibility with specific headgear systems.
  • Mesh screen
  • Black color
  • Measures 6 in. × 15-1/2 in.
  • Epoxy coating
  • No lens shade
  • Compatible with T-Lock Ratchet Style Headgear and Woodsman System