Learn More
Medchemexpress LLC 1H-pyrazole-1-acetamide, 4-[5,6-dihydro-2-[3-methyl-1-(1-methylethyl)-1H-1,2,4-triazol-5-yl]imidazo | 1282512-48-4 | MFCD26142641 | 99.8% | 460.53 g·mol⁻¹ | C24H28N8O2 | 5 MG
SDP
Catalog No. 501872983
Encompass_Preferred
Taselisib (GDC-0032) is a potent small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) used in preclinical research to probe PI3K signaling and PIK3CA-driven pathways. It displays high biochemical potency across PI3K isoforms and is provided as a research-grade compound for in vitro and in vivo studies.
  • Potent inhibition of PI3Kα with Ki ≈ 0.29 nM.
  • Sub-nanomolar activity versus PI3Kδ and PI3Kγ; low-nanomolar versus PI3Kβ.
  • High purity suitable for biological assays (99.8% LCMS).
  • Stable powder with recommended storage at -20°C; solutions stable at -80°C.
  • Molecular weight 460.53 g·mol⁻¹; chemical formula C24H28N8O2.
Catalog No. 50-187-2983 Supplier Medchemexpress LLC Supplier No. HY138985MG
Taselisib (GDC-0032) is a potent small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) used in preclinical research to probe PI3K signaling and PIK3CA-driven pathways. It displays high biochemical potency across PI3K isoforms and is provided as a research-grade compound for in vitro and in vivo studies.
  • Potent inhibition of PI3Kα with Ki ≈ 0.29 nM.
  • Sub-nanomolar activity versus PI3Kδ and PI3Kγ; low-nanomolar versus PI3Kβ.
  • High purity suitable for biological assays (99.8% LCMS).
  • Stable powder with recommended storage at -20°C; solutions stable at -80°C.
  • Molecular weight 460.53 g·mol⁻¹; chemical formula C24H28N8O2.