Learn More
Cayman Chemical Tasimelteon, Purity: ≥98%, CAS: 609799-22-6, 50 mg
SDP
Catalog No. 502995137
Encompass_Preferred
An MT receptor antagonist (Kis = 0.304 and 0.069 nM for MT1 and MT2, respectively, in NIH3T3 cells expressing the human recombinant receptors); selective for MT1 and MT2 receptors over a panel of 160 additional receptors and enzymes at 10 µM; inhibits forskolin-induced cAMP accumulation (EC50s = 0.79 and 1 nM, respectively) in NIH3T3 cells expressing the MT1 or MT2 receptor, respectively
Catalog No. 50-299-5137 Supplier Cayman Chemical Supplier No. 2354650 MG
An MT receptor antagonist (Kis = 0.304 and 0.069 nM for MT1 and MT2, respectively, in NIH3T3 cells expressing the human recombinant receptors); selective for MT1 and MT2 receptors over a panel of 160 additional receptors and enzymes at 10 µM; inhibits forskolin-induced cAMP accumulation (EC50s = 0.79 and 1 nM, respectively) in NIH3T3 cells expressing the MT1 or MT2 receptor, respectively