Learn More
Medchemexpress LLC Tasipimidine sulfate | 1465908-73-9 | 99.3% | 330.36 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY109075A10MM/1ML
Encompass_Preferred
Tasipimidine sulfate is an orally active and selective α2A-adrenoceptor agonist with a pEC50 of 7.57 for human α2A-adrenoceptors and an EC50 of 5.7 nM for rat α2-adrenoceptor. It can be used in research related to situational anxiety and fear.
  • Orally active and selective α2A-adrenoceptor agonist.
  • Reduces spontaneous locomotor activity in mice.
  • Reduces the amplitude of the acoustic startle reflex in rats.
  • Improves separation anxiety in dogs, leading to more positive owner ratings and significantly reduced destructive/rearranging behavior and vocalization at a dose of 30 μg/kg.
Catalog No. 50-003-45071