Learn More
Medchemexpress LLC Tasipimidine sulfate | 1465908-73-9 | 99.3% | 330.36 | 25 MG
SDP
Catalog No. 5000347598
Encompass_Preferred
Tasipimidine sulfate is an orally active and selective α2A-adrenoceptor agonist, demonstrating a pEC50 of 7.57 for human α2A-adrenoceptors and an EC50 of 5.7 nM for rat α2-adrenoceptor. This compound is suitable for research related to situational anxiety and fear, offering high purity and stability for laboratory applications.
  • Selected for α2A-adrenoceptors
  • Suitable for research on situational anxiety and fear
  • Reduces spontaneous locomotor activity in mice
  • Reduces acoustic startle reflex in rats
  • Improves separation anxiety in dogs
Catalog No. 50-003-47598 Supplier Medchemexpress LLC Supplier No. 25MGHY109075A
Tasipimidine sulfate is an orally active and selective α2A-adrenoceptor agonist, demonstrating a pEC50 of 7.57 for human α2A-adrenoceptors and an EC50 of 5.7 nM for rat α2-adrenoceptor. This compound is suitable for research related to situational anxiety and fear, offering high purity and stability for laboratory applications.
  • Selected for α2A-adrenoceptors
  • Suitable for research on situational anxiety and fear
  • Reduces spontaneous locomotor activity in mice
  • Reduces acoustic startle reflex in rats
  • Improves separation anxiety in dogs